Gene Symbol: Cypt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173412.2 | NP_775588.2 | 1 | 587 | ||
GenBank mRNA | AF463501.1 | - | 1 | 587 | ||
AK005791.1 | - | 1 | 580 | |||
BC048530.1 | - | 1 | 576 | |||
BC060952.1 | - | 1 | 576 | 6772242 |