Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115130.1 | NP_001108602.1 | 4 | 1606 | ||
NM_172765.3 | NP_766353.2 | 4 | 1552 | |||
GenBank mRNA | AK036895.1 | - | 4 | 1552 | ||
AV279920.2 | - | 3 | 461 | |||
BC037671.1 | - | 3 | 436 |