Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK031141.1 | - | 12 | 1249 | ||
AK158516.1 | - | 10 | 1064 | |||
AK161676.1 | - | 11 | 1192 |