Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039519.2 | NP_001034608.1 | 4 | 417 | ||
NM_001282083.1 | NP_001269012.1 | 4 | 708 | |||
GenBank mRNA | AK050621.1 | - | 4 | 686 | ||
AK164255.1 | - | 5 | 850 | |||
AK165466.1 | - | 4 | 415 | |||
AK169778.1 | - | 4 | 384 | |||
BC028551.1 | - | 5 | 538 | 1495874 | ||
BC048705.1 | - | 4 | 623 | 6505667 | ||
BG088284.2 | - | 4 | 376 |