Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153459.4 | NP_703189.3 | 3 | 1229 | ||
GenBank mRNA | AK140372.1 | - | 3 | 1230 | ||
AK158817.1 | - | 2 | 405 | |||
BC010207.1 | - | 3 | 872 | 3964527 | ||
BC025048.1 | - | 2 | 220 |