Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ago1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153403.2 | NP_700452.2 | 11 | 1331 | ||
GenBank mRNA | AB081471.1 | - | 11 | 1331 | ||
AK080954.1 | - | 9 | 1181 | |||
BC129914.1 | - | 11 | 1331 | 40111808 | ||
BC129915.1 | - | 11 | 1327 |