Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adgrg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079847.2 | NP_001073316.1 | 24 | 2511 | ||
NM_001079848.2 | NP_001073317.1 | 23 | 2481 | |||
NM_001079857.2 | NP_001073326.1 | 24 | 2520 | |||
NM_001290445.1 | NP_001277374.1 | 24 | 2514 | |||
NM_001290446.1 | NP_001277375.1 | 25 | 2553 | |||
NM_178712.4 | NP_848827.1 | 25 | 2562 | |||
GenBank mRNA | AF538952.1 | - | 25 | 2527 | ||
AF538955.1 | - | 24 | 2479 | |||
AF538956.1 | - | 25 | 2518 | |||
AF538957.1 | - | 23 | 2446 | |||
AK041291.1 | - | 26 | 2751 | |||
AK046871.1 | - | 24 | 2487 | |||
BC115874.1 | - | 23 | 2456 | 40086356 | ||
BC116644.1 | - | 24 | 2507 | 40086357 |
Set Membership: |
![]() |