Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Eml6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146016.2 | NP_666128.2 | 40 | 6403 | ||
GenBank mRNA | AK081418.1 | - | 5 | 754 | ||
BC024726.1 | - | 12 | 1838 | 5362808 |