Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175490.4 | NP_780699.2 | 2 | 1418 | ||
GenBank mRNA | AK044553.1 | - | 1 | 885 | ||
AK141614.1 | - | 2 | 1419 | |||
AY253852.1 | - | 1 | 1125 | |||
BC125614.1 | - | 1 | 1214 | 40130129 | ||
BC125616.1 | - | 1 | 1214 | 40130131 |
Set Membership: |
![]() |