Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arid4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081195.1 | NP_001074664.1 | 12, 13 | 1232 | ||
GenBank mRNA | AK050594.1 | - | 11, 12 | 1039 | ||
AK149171.1 | - | 2, 3 | 292 | |||
AY116674.1 | - | 12, 13 | 1118 | |||
BC025436.1 | - | 12, 13 | 1220 | |||
BC058209.1 | - | 12, 13 | 1149 | |||
BC080780.1 | - | 12, 13 | 1167 | |||
BC157975.1 | - | 11, 12 | 1034 | 9007297 | ||
BC158109.1 | - | 11, 12 | 1034 | 9007390 |