Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Synb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173420.3 | NP_775596.1 | 939 | |||
GenBank mRNA | AK086309.1 | - | 940 | |||
BC137938.1 | - | 681 | 8860957 | |||
BC137956.1 | - | 681 | 8860975 |