Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mief1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178719.5 | NP_848834.2 | 6 | 1452 | ||
GenBank mRNA | AK038466.1 | - | 6 | 1425 | ||
AK041184.1 | - | 6 | 1345 | |||
AK044773.1 | - | 7 | 1885 | |||
AK050546.1 | - | 6 | 1413 | |||
AK080375.1 | - | 6 | 1398 | |||
AK163389.1 | - | 7 | 1554 | |||
BC094597.1 | - | 6 | 1382 | |||
BC137633.1 | - | 5 | 1356 | 8860652 |