Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Liph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083894.1 | NP_001077363.1 | 2 | 513 | ||
NM_001289581.1 | NP_001276510.1 | 2 | 513 | |||
NM_001289582.1 | NP_001276511.1 | 2 | 513 | |||
NM_153404.3 | NP_700453.1 | 2 | 513 | |||
GenBank mRNA | AK048150.1 | - | 2 | 412 | ||
AK134352.1 | - | 2 | 472 | |||
AK136503.1 | - | 2 | 476 | |||
AK137278.1 | - | 2 | 485 | |||
AK161067.1 | - | 2 | 457 | |||
AK162492.1 | - | 2 | 479 | |||
AY093499.1 | - | 2 | 340 | |||
BC037489.1 | - | 2 | 431 | 4487939 |