Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc4a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271544.1 | NP_001258473.1 | 7 | 960 | ||
NM_001271545.1 | NP_001258474.1 | 7 | 960 | |||
NM_001271546.1 | NP_001258475.1 | 7 | 960 | |||
NM_001271547.1 | NP_001258476.1 | 7 | 960 | |||
NM_172830.2 | NP_766418.1 | 7 | 960 | |||
GenBank mRNA | AK050187.1 | - | 5 | 536 | ||
AK085269.1 | - | 7 | 960 | |||
AK085424.1 | - | 7 | 963 | |||
AK128930.1 | - | 7 | 962 | |||
AK142545.1 | - | 7 | 962 | |||
AK143939.1 | - | 7 | 968 | |||
BC113179.1 | - | 7 | 955 | 40060674 | ||
BC113785.1 | - | 7 | 955 |