Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175506.3 | NP_780715.1 | 14 | 2305 | ||
GenBank mRNA | AK029178.1 | - | 3 | 1638 | ||
AY135183.1 | - | 14 | 2305 | |||
BC150736.1 | - | 14 | 2402 | 9087647 |