Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ankmy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172850.3 | NP_766438.2 | 11 | 2186 | ||
GenBank mRNA | AK029700.1 | - | 11 | 2188 | ||
BC120887.1 | - | 10 | 2022 | 40060916 | ||
BC120888.1 | - | 10 | 2022 | 40060917 |