Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pin1rt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033768.2 | NP_001028940.1 | 2 | 490 | ||
GenBank mRNA | AK145372.1 | - | 2 | 491 | ||
BC147362.1 | - | 2 | 312 | 9056420 | ||
BC147363.1 | - | 2 | 312 | 9056439 |