Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2R2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206994.1 | NP_001193923.1 | 5 | 439 | ||
NM_001206995.1 | NP_001193924.1 | 5 | 543 | |||
NM_001206996.1 | NP_001193925.1 | 5 | 593 | |||
NM_020416.3 | NP_065149.2 | 4 | 731 | |||
NM_181876.2 | NP_870991.1 | 4 | 385 | |||
XM_011513495.1 | XP_011511797.1 | 4 | 480 | |||
XM_011513496.1 | XP_011511798.1 | 4 | 495 | |||
GenBank mRNA | AB209183.1 | - | 3 | 4007 | ||
AF086924.1 | - | 4 | 415 | |||
AF145027.1 | - | 4 | 578 | |||
AK093115.1 | - | 4 | 385 | |||
AK296474.1 | - | 5 | 439 | |||
AK316003.1 | - | 6 | 553 | |||
BC032954.2 | - | 5 | 593 | 4823280 | ||
BM551976.1 | - | 731 |