Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177732.4 | NP_808400.1 | 7, 8 | 658 | ||
GenBank mRNA | AK049180.1 | - | 7, 8 | 659 | ||
AK049462.1 | - | 7, 8 | 578 | |||
AK140305.1 | - | 7, 8 | 950 | |||
AK169775.1 | - | 7, 8 | 560 | |||
AK220330.1 | - | 7, 8 | 568 | |||
BC115984.1 | - | 8, 9 | 559 | 40105299 |