Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Zfp128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153802.4 | NP_722497.1 | 4 | 1645 | ||
GenBank mRNA | AF480861.1 | - | 4 | 1603 | ||
AK032046.1 | - | 4 | 1625 | |||
AK035906.1 | - | 4 | 1646 | |||
AK132244.1 | - | 3 | 1900 | |||
BC088998.1 | - | 4 | 1616 | 6417536 | ||
BC094248.1 | - | 4 | 1616 | 5718665 |