Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp473 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289836.1 | NP_001276765.1 | 5 | 3039 | ||
NM_001289837.1 | NP_001276766.1 | 5 | 3013 | |||
NM_001289838.1 | NP_001276767.1 | 5 | 2717 | |||
NM_001289839.1 | NP_001276768.1 | 5 | 2636 | |||
NM_178734.4 | NP_848849.2 | 5 | 2865 | |||
GenBank mRNA | AK030093.1 | - | 5 | 2865 | ||
AK036453.1 | - | 5 | 2675 | |||
AK050747.1 | - | 5 | 2713 | |||
AK173109.1 | - | 3 | 8284 | |||
BC096555.1 | - | 5 | 2634 | 30614720 |