Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ovch2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172908.3 | NP_766496.2 | 16 | 1755 | ||
GenBank mRNA | AK033776.1 | - | 15 | 1644 | ||
BC125284.1 | - | 16 | 1784 | 40129799 | ||
BC125288.1 | - | 16 | 1784 | 40129803 |