Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKAB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005399.4 | NP_005390.1 | 6 | 775 | ||
NR_103870.1 | 4 | 487 | ||||
NR_103871.1 | 5 | 608 | ||||
XM_011509729.1 | XP_011508031.1 | 6 | 775 | |||
XM_017001697.1 | XP_016857186.1 | 5 | 654 | |||
GenBank mRNA | AJ224538.1 | - | 5 | 591 | ||
AK292820.1 | - | 6 | 656 | |||
AK294863.1 | - | 5 | 580 | |||
AK304069.1 | - | 4 | 368 | |||
AK310982.1 | - | 6 | 656 | |||
AK316005.1 | - | 5 | 486 | |||
AK316183.1 | - | 4 | 459 | |||
BC053610.1 | - | 6 | 730 | 6091476 | ||
BU166852.1 | - | 2 | 92 | |||
KJ534919.1 | - | 5 | 591 | |||
KJ535024.1 | - | 4 | 424 | |||
KJ535025.1 | - | 5 | 591 |