Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173780.4 | NP_776141.1 | 6 | 961 | ||
GenBank mRNA | AK044185.1 | - | 6 | 962 | ||
BC070442.1 | - | 7 | 1248 | 30535488 |