Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atxn7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139227.4 | NP_631973.3 | 11 | 2321 | ||
GenBank mRNA | AF455111.1 | - | 10 | 2061 | ||
AK046679.1 | - | 11 | 2322 | |||
BC035459.1 | - | 10 | 1974 | |||
BC137846.1 | - | 11 | 2242 | 8860865 |