Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prokr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144944.3 | NP_659193.3 | 3 | 1199 | ||
GenBank mRNA | AF487279.1 | - | 2 | 877 | ||
AK030458.1 | - | 2 | 599 | |||
AK041586.1 | - | 3 | 1123 | |||
AK080980.1 | - | 3 | 1150 | |||
BC043116.1 | - | 3 | 1152 | 6408655 | ||
BC057557.1 | - | 3 | 1152 | 6408655 |
Set Membership: |
![]() |