Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhobtb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153514.5 | NP_705734.4 | 5 | 1394 | ||
GenBank mRNA | AF420001.1 | - | 4 | 885 | ||
AK037927.1 | - | 4 | 1921 | |||
AK038196.1 | - | 5 | 1486 | |||
AK081100.1 | - | 5 | 1323 | |||
AK122354.1 | - | 5 | 1375 | |||
AK171295.1 | - | 11 | 2009 | |||
BC026836.1 | - | 5 | 1375 | 4242008 | ||
BC056954.1 | - | 5 | 999 | 5685456 |