Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd300lf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169153.1 | NP_001162624.1 | 2 | 479 | ||
NM_145634.3 | NP_663609.2 | 2 | 479 | |||
GenBank mRNA | AF251703.1 | - | 2 | 433 | ||
AK089313.1 | - | 2 | 479 | |||
AK089465.1 | - | 2 | 478 | |||
AK154273.1 | - | 2 | 412 | |||
AK155367.1 | - | 2 | 477 | |||
AY214460.1 | - | 2 | 431 | |||
AY457047.1 | - | 2 | 368 | |||
BC057864.1 | - | 2 | 444 | 5318856 | ||
JX073136.1 | - | 2 | 454 |