Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKCQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242413.2 | NP_001229342.1 | 2 | 173 | ||
NM_001282644.1 | NP_001269573.1 | 2 | 173 | |||
NM_001282645.1 | NP_001269574.1 | 2 | 173 | |||
NM_001323265.1 | NP_001310194.1 | 2 | 228 | |||
NM_001323266.1 | NP_001310195.1 | 2 | 173 | |||
NM_001323267.1 | NP_001310196.1 | 2 | 173 | |||
NM_006257.4 | NP_006248.1 | 2 | 173 | |||
XM_005252496.4 | XP_005252553.1 | 2 | 273 | |||
XM_005252497.4 | XP_005252554.1 | 2 | 275 | |||
XM_006717465.3 | XP_006717528.1 | 4 | 2668 | |||
XM_017016410.1 | XP_016871899.1 | 3 | 2579 | |||
XM_017016411.1 | XP_016871900.1 | 4 | 2668 | |||
GenBank mRNA | AK293935.1 | - | 2 | 111 | ||
AK308099.1 | - | 2 | 148 | |||
AK312411.1 | - | 2 | 123 | |||
AU099414.1 | - | 2 | 148 | |||
AY702977.1 | - | 1 | 73 | |||
BC101465.1 | - | 1 | 92 | 8068971 | ||
BC113359.1 | - | 1 | 92 | 8322411 | ||
BX647657.1 | - | 1 | 257 | |||
L01087.1 | - | 2 | 167 | |||
L07032.1 | - | 1 | 94 |