Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr832 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011824.1 | NP_001011824.1 | 1 | 637 |
Gene Symbol: Olfr834 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011823.1 | NP_001011823.1 | 1 | 637 |