Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001292040.1 | NP_001278969.1 | 2 | 1526 | ||
NM_002747.3 | NP_002738.2 | 2 | 1526 | |||
XM_005258299.3 | XP_005258356.1 | 2 | 5888 | |||
XM_011526074.2 | XP_011524376.1 | 2 | 6070 | |||
XM_011526075.2 | XP_011524377.1 | 2 | 1528 | |||
XM_011526076.2 | XP_011524378.1 | 2 | 1891 | |||
XM_017025839.1 | XP_016881328.1 | 2 | 1459 | |||
GenBank mRNA | AK295058.1 | - | 2 | 1447 | ||
AK303601.1 | - | 1 | 188 | |||
BC040493.1 | - | 2 | 1498 | |||
BC050299.1 | - | 2 | 1526 | 5303135 | ||
BC117216.1 | - | 1 | 580 | 40125767 | ||
BC143769.1 | - | 1 | 578 | |||
BX648603.1 | - | 2 | 1515 | |||
X59727.1 | - | 1 | 1063 |