Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002751.6 | NP_002742.3 | 9, 10 | 866 | ||
NR_110887.1 | 9, 10 | 866 | ||||
XM_011530706.1 | XP_011529008.1 | 7, 8 | 590 | |||
GenBank mRNA | AK291845.1 | - | 9, 10 | 817 | ||
BC027933.1 | - | 9, 10 | 823 | 5199628 | ||
BX444192.2 | - | 9, 10 | 863 |