Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr509 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146372.1 | NP_666484.1 | 1 | 698 | ||
GenBank mRNA | BC132049.1 | - | 1 | 782 | 40130326 | |
BC132435.1 | - | 1 | 782 | 40130712 |
Set Membership: |
G-protein coupled receptor |