Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr695 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146598.2 | NP_666809.1 | 3 | 656 | ||
GenBank mRNA | BC104092.1 | - | 1 | 403 | 40042972 | |
BC104093.1 | - | 1 | 403 | 40042973 | ||
BC129798.1 | - | 1 | 403 | 40042986 |
Gene Symbol: Olfr694 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146452.2 | NP_666663.2 | 1 | 403 | ||
GenBank mRNA | BC129796.1 | - | 1 | 403 | 40042951 | |
BC129797.1 | - | 1 | 403 | 40042952 |