Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1261 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146474.1 | NP_666685.1 | 1 | 793 | ||
GenBank mRNA | AB076075.1 | - | 1 | 793 | ||
BC119116.1 | - | 1 | 825 | 8733865 | ||
BC119144.1 | - | 1 | 825 | 8733893 |
Gene Symbol: Olfr1263 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146794.1 | NP_667005.1 | 1 | 793 | ||
GenBank mRNA | BC119193.1 | - | 1 | 834 | 8733942 | |
BC119197.1 | - | 1 | 834 | 8733946 |