Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Olfr373 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146539.2 | NP_666750.2 | 1 | 901 | ||
GenBank mRNA | BC106979.2 | - | 1 | 899 | ||
BC106980.1 | - | 1 | 898 | 40042607 | ||
BC119444.1 | - | 1 | 901 | |||
BC119445.2 | - | 1 | 901 | 40042612 |