Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1364 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146540.2 | NP_666751.2 | 1 | 668 | ||
GenBank mRNA | BC127986.1 | - | 1 | 672 | 40135567 |