Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr736 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146666.1 | NP_666877.1 | 1 | 65 | ||
GenBank mRNA | BC110467.2 | - | 1 | 65 | 40041632 | |
BC127601.1 | - | 1 | 65 | 40041634 | ||
BC129795.1 | - | 1 | 63 |
Set Membership: |
![]() |