Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr767 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146318.2 | NP_666430.2 | 1 | 266 | ||
GenBank mRNA | BC137767.1 | - | 1 | 362 | 8860786 |
Gene Symbol: Olfr765 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085477.1 | NP_001078946.1 | 1 | 266 |