Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1248 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146791.2 | NP_667002.2 | 1 | 293 | ||
GenBank mRNA | BC150718.1 | - | 1 | 295 | 9087629 |
Gene Symbol: Olfr1250 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146965.1 | NP_667176.1 | 1 | 238 | ||
GenBank mRNA | BC125323.1 | - | 1 | 332 | 40129838 | |
BC125349.1 | - | 1 | 332 | 40129864 |
Gene Symbol: Olfr1252 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207568.1 | NP_997451.1 | 1 | 238 | ||
GenBank mRNA | BC125497.1 | - | 1 | 335 | 40130012 | |
BC125499.1 | - | 1 | 335 | 40130014 |