- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: Olfr1414 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147039.2 | NP_667250.1 | 2 | 1093 | ||
GenBank mRNA | BC120830.1 | - | 1 | 920 | 40129753 |