Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr572 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147089.2 | NP_667300.1 | 1 | 69 | ||
GenBank mRNA | BC128002.1 | - | 1 | 73 | 40135902 |