Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr559 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147112.1 | NP_667323.1 | 1 | 658 | ||
GenBank mRNA | BC119185.1 | - | 1 | 680 | 8733934 | |
BC120640.1 | - | 1 | 680 | 40129563 |
Gene Symbol: Olfr33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147073.1 | NP_667284.1 | 1 | 658 | ||
GenBank mRNA | BC104110.1 | - | 1 | 658 | 40043836 | |
BC104111.1 | - | 1 | 658 | 40043837 | ||
X89685.1 | - | 1 | 266 |