Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3pxd2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177364.3 | NP_796338.2 | 7 | 543 | ||
GenBank mRNA | AB430861.1 | - | 7 | 465 | ||
AK089330.1 | - | 7 | 515 | |||
AK090119.1 | - | 7 | 536 | |||
AK154898.1 | - | 7 | 501 | |||
AK163471.1 | - | 7 | 515 | |||
AK169978.1 | - | 7 | 535 | |||
AK171384.1 | - | 7 | 515 | |||
BC115711.1 | - | 7 | 512 | |||
BC115764.1 | - | 7 | 512 | |||
BC141306.1 | - | 7 | 515 | 9055918 |