Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zkscan17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130529.2 | NP_001124001.1 | 10 | 2128 | ||
NM_001291014.1 | NP_001277943.1 | 10 | 1882 | |||
NM_172941.4 | NP_766529.3 | 10 | 2175 | |||
GenBank mRNA | AK051711.1 | - | 10 | 1464 | ||
AK155196.1 | - | 10 | 1705 | |||
AK155490.1 | - | 10 | 1757 | |||
AK170924.1 | - | 10 | 1705 | |||
AK170989.1 | - | 10 | 1710 | |||
AK171190.1 | - | 10 | 1757 | |||
AK183250.1 | - | 1 | 141 | |||
AY302242.1 | - | 9 | 1656 | |||
BC040205.1 | - | 9 | 1660 | 4947290 |