Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgat5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172948.3 | NP_766536.2 | 4 | 1046 | ||
GenBank mRNA | AB116028.1 | - | 4 | 433 | ||
AB119127.1 | - | 4 | 512 | |||
BC063084.1 | - | 4 | 1046 | |||
BC094604.1 | - | 4 | 1046 | 6822690 |