Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fam107a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183187.3 | NP_899010.1 | 2 | 372 | ||
GenBank mRNA | AK044219.1 | - | 2 | 142 | ||
AK165722.1 | - | 3 | 559 | |||
BC055107.1 | - | 2 | 372 | 6532624 |