Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145921.1 | NP_001139393.1 | 18 | 4044 | ||
NM_001145922.1 | NP_001139394.1 | 18 | 4044 | |||
NM_198249.4 | NP_937892.2 | 18 | 4044 | |||
GenBank mRNA | AK131136.1 | - | 18 | 4013 | ||
AK143527.1 | - | 18 | 4019 | |||
AK163945.1 | - | 18 | 4237 | |||
BC030925.1 | - | 5 | 1203 | |||
BC058405.1 | - | 4 | 503 | |||
BC059018.1 | - | 18 | 3946 | 6848691 |