Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stfa2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173869.3 | NP_776294.1 | 3 | 290 | ||
GenBank mRNA | AK050597.1 | - | 3 | 291 | ||
AY163161.1 | - | 3 | 266 | |||
BC117052.1 | - | 3 | 288 | 40126371 | ||
BC119354.1 | - | 3 | 288 | 8734103 |
Gene Symbol: Stfa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082545.1 | NP_001076014.1 | 3 | 294 | ||
GenBank mRNA | BC138004.1 | - | 3 | 388 | 8861023 | |
M92418.1 | - | 3 | 266 |