Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270482.1 | NP_001257411.1 | 12 | 1173 | ||
NM_002815.3 | NP_002806.2 | 12 | 1173 | |||
GenBank mRNA | AB003102.1 | - | 12 | 1123 | ||
AF001212.1 | - | 12 | 1152 | |||
AK093602.1 | - | 2 | 1837 | |||
AK223196.1 | - | 12 | 1126 | |||
AK290602.1 | - | 12 | 1162 | |||
BC000437.2 | - | 12 | 1120 | 2820583 | ||
BC004430.1 | - | 12 | 1120 | 2820583 |